Assay Details
| Gene Symbol: FBXL21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270994.1 | NP_001257923.1 | 7 - 8 | 1295 | 83 | |
| NM_001270995.1 | NP_001257924.1 | 6 - 7 | 1118 | 83 | ||
| XM_015293970.2 | XP_015149456.1 | 6 - 7 | 1131 | 83 | ||
| XM_015293971.2 | XP_015149457.1 | 5 - 6 | 992 | 83 | ||
| XM_025154853.1 | XP_025010621.1 | 6 - 7 | 1144 | 83 | ||
| GenBank mRNA | BU268472.1 | - | 3 - 4 | 495 | 83 | |
| BX270427.4 | - | 1 - 2 | 146 | 83 | ||
More Information
Set Membership: |
|

FBXL21
Genomic Map