Assay Details
| Gene Symbol: TIMM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271154.1 | NP_001258083.1 | 3 - 4 | 210 | 196 | |
| NM_001271155.1 | NP_001258084.1 | 3 - 4 | 322 | 196 | ||
| XM_015287252.2 | XP_015142738.1 | 3 - 4 | 206 | 196 | ||
| XM_015287253.2 | XP_015142739.1 | 3 - 4 | 264 | 196 | ||
| GenBank mRNA | AJ720716.1 | - | 3 - 4 | 203 | 196 | |
| DQ066930.1 | - | 2 - 3 | 179 | 196 | ||
More Information
Set Membership: |
|

TIMM9
Genomic Map