Assay Details
| Gene Symbol: TRAPPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271159.1 | NP_001258088.1 | 1 - 2 | 47 | 54 | |
| GenBank mRNA | AJ851791.1 | - | 1 - 2 | 47 | 54 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ851791.1, NM_001271159.1 transcripts.
Set Membership: |
|

TRAPPC5
Genomic Map