Assay Details
| Gene Symbol: ISCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271936.1 | NP_001258865.1 | 1 - 2 | 172 | 96 | |
| GenBank mRNA | AJ720560.1 | - | 1 - 2 | 131 | 96 | |
| BU398918.1 | - | 1 - 2 | 172 | 96 | ||
More Information
Set Membership: |
|

ISCA1
Genomic Map