Assay Details
| Gene Symbol: ATG4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271986.1 | NP_001258915.1 | 3 - 4 | 318 | 72 | |
| NM_001271987.1 | NP_001258916.1 | 3 - 4 | 267 | 72 | ||
| XM_015278325.2 | XP_015133811.1 | 3 - 4 | 224 | 72 | ||
| GenBank mRNA | AJ720667.1 | - | 3 - 4 | 273 | 72 | |
| CF256828.1 | - | 3 - 4 | 223 | 72 | ||
More Information
Set Membership: |
|

ATG4A
Genomic Map