Assay Details
| Gene Symbol: SMIM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271991.1 | NP_001258920.1 | 1 | 917 | 86 | |
| GenBank mRNA | CD736767.1 | - | 1 | 263 | 86 | |
| CR353728.1 | - | 1 | 836 | 86 | ||
| CR387775.1 | - | 2 | 707 | 86 | ||
More Information
Set Membership: |
|

SMIM12
Genomic Map