Assay Details
| Gene Symbol: PTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276362.1 | NP_001263291.1 | 1 - 2 | 304 | 64 | |
| GenBank mRNA | BU351780.1 | - | 1 - 2 | 208 | 64 | |
| BU388475.1 | - | 1 - 2 | 304 | 64 | ||
| BU400419.1 | - | 1 - 2 | 149 | 64 | ||
| BU441781.1 | - | 1 - 2 | 43 | 64 | ||
| CN217783.1 | - | 1 - 2 | 144 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001276362.1 transcripts.
Set Membership: |
|

PTN
Genomic Map