Assay Details
| Gene Symbol: TMEM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276388.1 | NP_001263317.1 | 3 - 4 | 402 | 64 | |
| NR_075086.1 | 4 - 5 | 571 | 64 | |||
| GenBank mRNA | BU384987.1 | - | 3 - 4 | 338 | 64 | |
| BU455881.1 | - | 3 - 4 | 336 | 64 | ||
| CN231678.1 | - | 3 - 4 | 331 | 64 | ||
More Information
Set Membership: |
|

TMEM17
Genomic Map