Assay Details
| Gene Symbol: BHLHE40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277144.2 | NP_001264073.1 | 2 - 3 | 296 | 94 | |
| GenBank mRNA | AM065613.1 | - | 2 - 3 | 283 | 94 | |
| BX930095.1 | - | 2 - 3 | 256 | 94 | ||
| BX930582.1 | - | 2 - 3 | 250 | 94 | ||
| BX931163.1 | - | 2 - 3 | 246 | 94 | ||
| DR424259.1 | - | 2 - 3 | 239 | 94 | ||
More Information
Set Membership: |
|

BHLHE40
Genomic Map