Assay Details
| Gene Symbol: ENY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277432.1 | NP_001264361.1 | 3 - 4 | 413 | 101 | |
| NM_001277433.1 | NP_001264362.1 | 4 - 5 | 276 | 101 | ||
| XM_025147116.1 | XP_025002884.1 | 4 - 5 | 341 | 101 | ||
| GenBank mRNA | BU259725.1 | - | 4 - 5 | 270 | 101 | |
| BX258120.3 | - | 3 - 4 | 236 | 101 | ||
| BX272505.4 | - | 3 - 4 | 240 | 101 | ||
| CF252535.1 | - | 4 - 5 | 262 | 101 | ||
| DN930008.1 | - | 3 - 4 | 413 | 101 | ||
More Information
Set Membership: |
|

ENY2
Genomic Map