Assay Details
| Gene Symbol: MED30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277436.1 | NP_001264365.1 | 2 - 3 | 396 | 111 | |
| NM_001277438.1 | NP_001264367.1 | 2 - 3 | 297 | 111 | ||
| XM_015283022.2 | XP_015138508.1 | 2 - 3 | 259 | 111 | ||
| XM_025147120.1 | XP_025002888.1 | 2 - 3 | 243 | 111 | ||
| GenBank mRNA | BX930101.1 | - | 2 - 3 | 287 | 111 | |
| BX932068.1 | - | 2 - 3 | 327 | 111 | ||
| CR406080.1 | - | 2 - 3 | 297 | 111 | ||
More Information
Set Membership: |
|

MED30
Genomic Map