Assay Details
| Gene Symbol: LXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277488.1 | NP_001264417.1 | 4 - 5 | 524 | 102 | |
| XM_015291694.1 | XP_015147180.1 | 3 - 4 | 1028 | 102 | ||
| GenBank mRNA | BU454341.1 | - | 1 - 2 | 68 | 102 | |
| BX929401.2 | - | 4 - 5 | 495 | 102 | ||
More Information
Set Membership: |
|

LXN
Genomic Map