Assay Details
| Gene Symbol: HPD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277491.1 | NP_001264420.1 | 10 - 11 | 840 | 53 | |
| XM_025155458.1 | XP_025011226.1 | 10 - 11 | 999 | 53 | ||
| GenBank mRNA | BX934087.1 | - | 10 - 11 | 840 | 53 | |
| BX935030.2 | - | 11 - 12 | 843 | 53 | ||
More Information
Set Membership: |
|

HPD
Genomic Map