Assay Details
| Gene Symbol: CDIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277612.1 | NP_001264541.1 | 5 - 6 | 981 | 98 | |
| XM_015294236.2 | XP_015149722.1 | 5 - 6 | 646 | 98 | ||
| XM_015294237.2 | XP_015149723.1 | 5 - 6 | 650 | 98 | ||
| XM_015294238.2 | XP_015149724.1 | 5 - 6 | 740 | 98 | ||
| XM_015294239.2 | XP_015149725.1 | 5 - 6 | 736 | 98 | ||
| XM_025155112.1 | XP_025010880.1 | 5 - 6 | 738 | 98 | ||
| XM_025155113.1 | XP_025010881.1 | 5 - 6 | 735 | 98 | ||
| GenBank mRNA | DR426031.1 | - | 5 - 6 | 660 | 98 | |
More Information
Set Membership: |
|

CDIP1
Genomic Map