Assay Details
| Gene Symbol: CALML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277629.1 | NP_001264558.1 | 3 - 4 | 215 | 172 | |
| GenBank mRNA | BX933823.1 | - | 3 - 4 | 230 | 172 | |
| BX933828.2 | - | 3 - 4 | 220 | 172 | ||
| BX935760.2 | - | 2 - 3 | 278 | 172 | ||
More Information
Set Membership: |
|

CALML4
Genomic Map