Assay Details
| Gene Symbol: TACSTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277676.1 | NP_001264605.1 | 1 | 525 | 110 | |
| GenBank mRNA | CF256804.1 | - | 1 | 525 | 110 | |
More Information
Set Membership: |
|

TACSTD2
Genomic Map