Assay Details
| Gene Symbol: RAB24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277713.1 | NP_001264642.1 | 4 - 5 | 366 | 58 | |
| GenBank mRNA | BX929971.2 | - | 4 - 5 | 366 | 58 | |
| BX935765.1 | - | 4 - 5 | 324 | 58 | ||
More Information
Set Membership: |
|

RAB24
Genomic Map