Assay Details
| Gene Symbol: CERS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277765.1 | NP_001264694.1 | 5 - 6 | 872 | 111 | |
| GenBank mRNA | CD218363.1 | - | 2 - 3 | 283 | 111 | |
More Information
Set Membership: |
|

CERS1
Genomic Map