Assay Details
Gene Symbol: EFCAB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277774.1 | NP_001264703.1 | 7 - 8 | 945 | 87 | |
XM_015291009.2 | XP_015146495.1 | 7 - 8 | 1137 | 87 | ||
GenBank mRNA | BU411059.1 | - | 4 - 5 | 462 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
