Assay Details
| Gene Symbol: PLAC8L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277782.1 | NP_001264711.1 | 3 - 4 | 286 | 62 | |
| XM_015276323.1 | XP_015131809.1 | 3 - 4 | 330 | 62 | ||
| XM_015276324.2 | XP_015131810.1 | 3 - 4 | 291 | 62 | ||
| XM_025149758.1 | XP_025005526.1 | 6 - 7 | 848 | 62 | ||
| GenBank mRNA | BX263772.4 | - | 2 - 3 | 268 | 62 | |
| CF250258.1 | - | 3 - 4 | 288 | 62 | ||
More Information
Set Membership: |
|

PLAC8L1
Genomic Map