Assay Details
| Gene Symbol: PLAC8L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277782.1 | NP_001264711.1 | 4 - 5 | 396 | 182 | |
| XM_015276323.1 | XP_015131809.1 | 4 - 5 | 440 | 182 | ||
| XM_015276324.2 | XP_015131810.1 | 4 - 5 | 401 | 182 | ||
| XM_025149758.1 | XP_025005526.1 | 7 - 8 | 958 | 182 | ||
| GenBank mRNA | BX263772.4 | - | 3 - 4 | 378 | 182 | |
| CF250258.1 | - | 4 - 5 | 398 | 182 | ||
More Information
Set Membership: |
|

PLAC8L1
Genomic Map