Assay Details
| Gene Symbol: IMMP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277878.1 | NP_001264807.1 | 4 - 5 | 528 | 80 | |
| XM_025150845.1 | XP_025006613.1 | 4 - 5 | 388 | 80 | ||
| GenBank mRNA | BX934258.1 | - | 1 - 2 | 108 | 80 | |
| BX950753.1 | - | 4 - 5 | 381 | 80 | ||
More Information
Set Membership: |
|

IMMP1L
Genomic Map