Assay Details
| Gene Symbol: GRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277900.1 | NP_001264829.1 | 2 - 3 | 559 | 113 | |
| GenBank mRNA | BX930380.2 | - | 2 - 3 | 532 | 113 | |
| BX935663.1 | - | 2 - 3 | 559 | 113 | ||
More Information
Set Membership: |
|

GRP
Genomic Map