Assay Details
| Gene Symbol: SNRPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277998.1 | NP_001264927.1 | 1 - 2 | 84 | 107 | |
| XM_015281596.2 | XP_015137082.1 | 2 - 3 | 272 | 107 | ||
| GenBank mRNA | BX930256.1 | - | 2 - 3 | 272 | 107 | |
| BX932288.2 | - | 2 - 3 | 287 | 107 | ||
More Information
Set Membership: |
|

SNRPD1
Genomic Map