Assay Details
| Gene Symbol: MRPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278026.1 | NP_001264955.1 | 1 - 2 | 102 | 69 | |
| NM_001278027.1 | NP_001264956.1 | 1 - 2 | 102 | 69 | ||
| NM_001278028.1 | NP_001264957.1 | 1 - 2 | 102 | 69 | ||
| GenBank mRNA | BU478730.1 | - | 1 - 2 | 104 | 69 | |
| BX931807.1 | - | 1 - 2 | 44 | 69 | ||
| CR352581.1 | - | 1 - 2 | 105 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278026.1, NM_001278027.1, NM_001278028.1 transcripts.
Set Membership: |
|

MRPS16
Genomic Map