Assay Details
| Gene Symbol: MRPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278026.1 | NP_001264955.1 | 2 - 3 | 369 | 84 | |
| NM_001278027.1 | NP_001264956.1 | 2 - 3 | 369 | 84 | ||
| GenBank mRNA | BU225343.1 | - | 2 - 3 | 290 | 84 | |
| BU478730.1 | - | 2 - 3 | 371 | 84 | ||
| BX274280.4 | - | 2 - 3 | 288 | 84 | ||
| BX929710.2 | - | 1 - 2 | 365 | 84 | ||
| BX931366.1 | - | 2 - 3 | 297 | 84 | ||
| BX931807.1 | - | 2 - 3 | 311 | 84 | ||
| CR352581.1 | - | 2 - 3 | 372 | 84 | ||
| CR405788.1 | - | 1 - 2 | 366 | 84 | ||
More Information
Set Membership: |
|

MRPS16
Genomic Map