Assay Details
| Gene Symbol: ITGB1BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278034.1 | NP_001264963.1 | 2 - 3 | 280 | 104 | |
| NM_001278035.1 | NP_001264964.1 | 2 - 3 | 280 | 104 | ||
| NM_001278036.1 | NP_001264965.1 | 2 - 3 | 280 | 104 | ||
| NM_001278038.1 | NP_001264967.1 | 3 - 4 | 435 | 104 | ||
| NM_001278052.1 | NP_001264981.1 | 2 - 3 | 280 | 104 | ||
| NR_102745.1 | 3 - 4 | 318 | 104 | |||
| XM_015276030.1 | XP_015131516.1 | 3 - 4 | 780 | 104 | ||
| XM_015276031.2 | XP_015131517.1 | 3 - 4 | 489 | 104 | ||
| XM_015276033.1 | XP_015131519.1 | 3 - 4 | 209 | 104 | ||
| XM_015276036.2 | XP_015131522.1 | 3 - 4 | 227 | 104 | ||
| XM_025148774.1 | XP_025004542.1 | 3 - 4 | 234 | 104 | ||
| XM_025148775.1 | XP_025004543.1 | 3 - 4 | 283 | 104 | ||
| XM_025148776.1 | XP_025004544.1 | 2 - 3 | 339 | 104 | ||
| XM_025148777.1 | XP_025004545.1 | 2 - 3 | 1063 | 104 | ||
| XM_025148778.1 | XP_025004546.1 | 4 - 5 | 359 | 104 | ||
| GenBank mRNA | BU144417.1 | - | 3 - 4 | 444 | 104 | |
| BU367456.1 | - | 2 - 3 | 217 | 104 | ||
| BU458846.1 | - | 3 - 4 | 209 | 104 | ||
| BX950700.1 | - | 2 - 3 | 136 | 104 | ||
| CD738050.1 | - | 2 - 3 | 208 | 104 | ||
| CR406232.1 | - | 2 - 3 | 178 | 104 | ||
| CV854210.1 | - | 2 - 3 | 251 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278038.1, NM_001278052.1, XM_015276030.1, XM_015276031.2, XM_015276033.1, XM_025148777.1, XM_025148778.1 transcripts.
Set Membership: |
|

ITGB1BP1
Genomic Map