Assay Details
| Gene Symbol: CBY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278149.1 | NP_001265078.1 | 2 - 3 | 517 | 84 | |
| GenBank mRNA | BX931504.2 | - | 2 - 3 | 516 | 84 | |
| BX931738.2 | - | 2 - 3 | 358 | 84 | ||
| CR406061.1 | - | 2 - 3 | 337 | 84 | ||
| CR407579.1 | - | 2 - 3 | 381 | 84 | ||
More Information
Set Membership: |
|

CBY1
Genomic Map