Assay Details
| Gene Symbol: JMJD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278726.1 | NP_001265655.1 | 6 - 7 | 504 | 64 | |
| GenBank mRNA | BU248795.1 | - | 6 - 7 | 438 | 64 | |
| CR390235.1 | - | 4 - 5 | 441 | 64 | ||
More Information
Set Membership: |
|

JMJD8
Genomic Map