Assay Details
| Gene Symbol: PAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281743.1 | NP_001268672.1 | 4 - 5 | 812 | 66 | |
| NM_001281745.1 | NP_001268674.1 | 4 - 5 | 602 | 66 | ||
More Information
Set Membership: |
|

PAIP1
Genomic Map