Assay Details
| Gene Symbol: C15orf61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282329.1 | NP_001269258.1 | 1 - 2 | 355 | 64 | |
| GenBank mRNA | BM490311.1 | - | 1 - 2 | 345 | 64 | |
| BU276799.1 | - | 1 - 2 | 267 | 64 | ||
| BX258038.4 | - | 1 - 2 | 99 | 64 | ||
More Information
Set Membership: |
|

C15orf61
Genomic Map