Assay Details
| Gene Symbol: GEMIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282339.1 | NP_001269268.1 | 1 | 2919 | 99 | |
| GenBank mRNA | AJ851571.1 | - | 1 | 2920 | 99 | |
More Information
Set Membership: |
|

GEMIN4
Genomic Map