Assay Details
| Gene Symbol: FAXDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285883.1 | NP_001272812.1 | 9 - 10 | 1194 | 61 | |
| XM_015293773.2 | XP_015149259.1 | 10 - 11 | 2325 | 61 | ||
| XM_015293774.2 | XP_015149260.1 | 10 - 11 | 2852 | 61 | ||
| XM_015293776.2 | XP_015149262.1 | 10 - 11 | 3092 | 61 | ||
| XM_015293777.2 | XP_015149263.1 | 10 - 11 | 3101 | 61 | ||
| XM_015293778.2 | XP_015149264.1 | 10 - 11 | 3076 | 61 | ||
| XM_015293779.2 | XP_015149265.1 | 10 - 11 | 3067 | 61 | ||
| XM_015293780.2 | XP_015149266.1 | 9 - 10 | 2879 | 61 | ||
| XM_025154848.1 | XP_025010616.1 | 8 - 9 | 1276 | 61 | ||
| XM_025154849.1 | XP_025010617.1 | 10 - 11 | 3000 | 61 | ||
| GenBank mRNA | BX262507.4 | - | 3 - 4 | 465 | 61 | |
More Information
Set Membership: |
|

FAXDC2
Genomic Map