Assay Details
| Gene Symbol: MARCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286224.1 | NP_001273153.1 | 2 - 3 | 203 | 69 | |
| XM_004940913.3 | XP_004940970.2 | 3 - 4 | 234 | 69 | ||
| XM_015285291.2 | XP_015140777.1 | 4 - 5 | 374 | 69 | ||
| XM_015285292.2 | XP_015140778.1 | 5 - 6 | 444 | 69 | ||
| XM_015285293.2 | XP_015140779.1 | 5 - 6 | 450 | 69 | ||
| XM_015285295.2 | XP_015140781.1 | 4 - 5 | 443 | 69 | ||
| XM_015285296.2 | XP_015140782.1 | 4 - 5 | 372 | 69 | ||
| XM_015285297.2 | XP_015140783.1 | 4 - 5 | 441 | 69 | ||
| XM_025149588.1 | XP_025005356.1 | 4 - 5 | 483 | 69 | ||
| XM_025149589.1 | XP_025005357.1 | 5 - 6 | 559 | 69 | ||
| XM_025149590.1 | XP_025005358.1 | 5 - 6 | 570 | 69 | ||
| XM_025149591.1 | XP_025005359.1 | 3 - 4 | 343 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004940913.3, XM_025149591.1 transcripts.
Set Membership: |
|

MARCH1
Genomic Map