Assay Details
| Gene Symbol: MARCH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286224.1 | NP_001273153.1 | 3 - 4 | 275 | 81 | |
| XM_015285296.2 | XP_015140782.1 | 5 - 6 | 444 | 81 | ||
| XM_015285297.2 | XP_015140783.1 | 5 - 6 | 513 | 81 | ||
More Information
Set Membership: |
|

MARCH1
Genomic Map