Assay Details
| Gene Symbol: NPRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287196.2 | NP_001274125.1 | 6 - 7 | 727 | 75 | |
| GenBank mRNA | BX934002.1 | - | 6 - 7 | 734 | 75 | |
More Information
Set Membership: |
|

NPRL2
Genomic Map