Assay Details
| Gene Symbol: RASA4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291781.1 | NP_001278710.1 | 10 - 11 | 1093 | 83 | |
| XM_025141941.1 | XP_024997709.1 | 10 - 11 | 1105 | 83 | ||
More Information
Set Membership: |
|

RASA4B
Genomic Map