Assay Details
| Gene Symbol: RASA4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291781.1 | NP_001278710.1 | 9 - 10 | 940 | 78 | |
| XM_025141941.1 | XP_024997709.1 | 9 - 10 | 952 | 78 | ||
More Information
Set Membership: |
|

RASA4B
Genomic Map