Assay Details
| Gene Symbol: LEO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001292086.3 | NP_001279015.2 | 7 - 8 | 1330 | 70 | |
| GenBank mRNA | CN232844.1 | - | 6 - 7 | 634 | 70 | |
More Information
Set Membership: |
|

LEO1
Genomic Map