Assay Details
| Gene Symbol: PTPN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293185.1 | NP_001280114.1 | 8 - 9 | 1057 | 56 | |
| GenBank mRNA | AJ393646.1 | - | 3 - 4 | 455 | 56 | |
More Information
Set Membership: |
|

PTPN9
Genomic Map