Assay Details
| Gene Symbol: HDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293288.1 | NP_001280217.1 | 11 - 12 | 1293 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HDC| Gene Symbol: HDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293288.1 | NP_001280217.1 | 11 - 12 | 1293 | 77 | |
Set Membership: |
|