Assay Details
| Gene Symbol: IPPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293322.1 | NP_001280251.1 | 3 - 4 | 320 | 82 | |
| XM_025154461.1 | XP_025010229.1 | 3 - 4 | 648 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025154461.1 transcripts.
Set Membership: |
|

IPPK
Genomic Map