Assay Details
| Gene Symbol: SRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301686.1 | NP_001288615.1 | 2 - 3 | 158 | 159 | |
| GenBank mRNA | AM068570.1 | - | 2 - 3 | 158 | 159 | |
| CR338823.1 | - | 2 - 3 | 173 | 159 | ||
| CR338992.2 | - | 2 - 3 | 163 | 159 | ||
More Information
Set Membership: |
|

SRA1
Genomic Map