Assay Details
Gene Symbol: C7orf73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302133.1 | NP_001289062.1 | 1 - 2 | 93 | 107 | |
GenBank mRNA | BX273924.4 | - | 1 - 2 | 93 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302133.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
