Assay Details
| Gene Symbol: NRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302155.1 | NP_001289084.1 | 2 - 3 | 369 | 160 | |
| GenBank mRNA | CN221487.1 | - | 2 - 3 | 273 | 160 | |
More Information
Set Membership: |
|

NRGN
Genomic Map