Assay Details
| Gene Symbol: S100Z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302198.1 | NP_001289127.1 | 2 - 3 | 482 | 92 | |
| XM_015277353.2 | XP_015132839.1 | 2 - 3 | 353 | 92 | ||
More Information
Set Membership: |
|

S100Z
Genomic Map