Assay Details
| Gene Symbol: GPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305060.2 | NP_001291989.1 | 3 - 4 | 697 | 60 | |
| GenBank mRNA | KF040585.1 | - | 3 - 4 | 697 | 60 | |
More Information
Set Membership: |
|

GPC1
Genomic Map