Assay Details
| Gene Symbol: MCM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305089.1 | NP_001292018.1 | 2 - 3 | 625 | 68 | |
| XM_015284462.2 | XP_015139948.1 | 3 - 4 | 737 | 68 | ||
| GenBank mRNA | AB689141.1 | - | 2 - 3 | 625 | 68 | |
More Information
Set Membership: |
|

MCM9
Genomic Map