Assay Details
| Gene Symbol: TMEM173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305152.1 | NP_001292081.1 | 3 - 4 | 865 | 76 | |
| XM_001232170.5 | XP_001232171.2 | 3 - 4 | 2860 | 76 | ||
| XM_015293523.2 | XP_015149009.1 | 4 - 5 | 2749 | 76 | ||
| XM_015293524.2 | XP_015149010.1 | 4 - 5 | 2277 | 76 | ||
| XM_015293525.2 | XP_015149011.1 | 4 - 5 | 2461 | 76 | ||
| XM_015293527.2 | XP_015149013.1 | 4 - 5 | 1964 | 76 | ||
| XM_015293528.2 | XP_015149014.1 | 3 - 4 | 2831 | 76 | ||
| XM_015293530.2 | XP_015149016.1 | 2 - 3 | 300 | 76 | ||
| XM_025154875.1 | XP_025010643.1 | 3 - 4 | 1029 | 76 | ||
| XM_025154876.1 | XP_025010644.1 | 3 - 4 | 1828 | 76 | ||
| XM_025154877.1 | XP_025010645.1 | 4 - 5 | 707 | 76 | ||
| GenBank mRNA | CD738576.1 | - | 2 - 3 | 363 | 76 | |
| KM879873.1 | - | 2 - 3 | 326 | 76 | ||
| KP893157.1 | - | 1 - 2 | 246 | 76 | ||
More Information
Set Membership: |
|

TMEM173
Genomic Map