Assay Details
| Gene Symbol: SLC31A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305656.1 | NP_001292585.1 | 5 - 6 | 637 | 103 | |
| NM_001305657.1 | NP_001292586.1 | 5 - 6 | 534 | 103 | ||
| NM_001305658.1 | NP_001292587.1 | 5 - 6 | 526 | 103 | ||
| NM_001305659.1 | NP_001292588.1 | 4 - 5 | 452 | 103 | ||
| NM_001305660.1 | NP_001292589.1 | 4 - 5 | 440 | 103 | ||
| XM_015279516.2 | XP_015135002.1 | 4 - 5 | 509 | 103 | ||
| XM_025156080.1 | XP_025011848.1 | 4 - 5 | 525 | 103 | ||
| XM_025156081.1 | XP_025011849.1 | 5 - 6 | 826 | 103 | ||
| XM_025156082.1 | XP_025011850.1 | 5 - 6 | 551 | 103 | ||
| XM_025156083.1 | XP_025011851.1 | 4 - 5 | 758 | 103 | ||
| XM_025156084.1 | XP_025011852.1 | 4 - 5 | 936 | 103 | ||
| GenBank mRNA | BX929472.1 | - | 5 - 6 | 523 | 103 | |
| BX929736.2 | - | 4 - 5 | 459 | 103 | ||
| BX934786.2 | - | 3 - 4 | 403 | 103 | ||
| BX935134.2 | - | 5 - 6 | 636 | 103 | ||
More Information
Set Membership: |
|

SLC31A1
Genomic Map